C1120

Sigma-Aldrich

CK2α1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):
CSNK2A1
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

91-123 nmol/min·mg

mol wt

~70 kDa

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSNK2A1(1457)

Biochem/physiol Actions

CK2a1 is a serine-threonine protein kinase whose targets include many critical regulators of cellular growth. It is highly expressed in a lymphopro-liferative disease of cattle and in many human cancers. Overexpression of the CK2 catalytic subunit in lymphocytes of transgenic mice leads to T cell lymphoma. The highest CK2a1 activity is found in mouse testicles and brain, followed by spleen, liver, lung, kidney and heart.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Sigma-Aldrich Co. LLC

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
B Guerra et al.
FEBS letters, 462(3), 353-357 (2000-01-06)
The highest CK2 activity was found in mouse testicles and brain, followed by spleen, liver, lung, kidney and heart. The activity values were directly correlated with the protein expression level of the CK2 subunits alpha (catalytic) and beta (regulatory). The...
I R Rifkin et al.
Journal of immunology (Baltimore, Md. : 1950), 161(10), 5164-5170 (1998-11-20)
MRL-lpr/lpr mice have a Fas receptor mutation that leads to abnormalities of apoptosis, lymphoproliferation, and a lupus-like autoimmune disease associated with the production of autoantibodies. Other than Fas pathway defects, little is known about molecular abnormalities that predispose to autoimmunity....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.