C212180

Sigma-Aldrich

DMT-2′O-Methyl-rC(tac) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):
N-[[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-cytidine, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-2′-O-Me-rC(tac) amidite
Empirical Formula (Hill Notation):
C52H64N5O10P
CAS Number:
Molecular Weight:
950.07
MDL number:
PubChem Substance ID:
NACRES:
NA.51

product line

Proligo Reagents

Quality Level

assay

≥98.0% (reversed phase HPLC)
≥99% (31P-NMR)

application(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for PerkinElmer
configured for Polygen

nucleoside profile

base: cytidine
base protecting group: TAC
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)O[C@H]1N5C=CC(NC(=O)COc6ccc(cc6)C(C)(C)C)=NC5=O

InChI

1S/C52H64N5O10P/c1-35(2)57(36(3)4)68(65-32-14-30-53)67-47-44(33-64-52(38-15-12-11-13-16-38,39-19-23-41(60-8)24-20-39)40-21-25-42(61-9)26-22-40)66-49(48(47)62-10)56-31-29-45(55-50(56)59)54-46(58)34-63-43-27-17-37(18-28-43)51(5,6)7/h11-13,15-29,31,35-36,44,47-49H,14,32-34H2,1-10H3,(H,54,55,58,59)/t44-,47-,48-,49-,68?/m1/s1

InChI key

JRJMGDLRRSEUHM-MVHRPHEDSA-N

Looking for similar products? Visit Product Comparison Guide

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service