Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C27H44O5
CAS Number:
Molecular Weight:
448.64
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
form
solid
SMILES string
[H]C12CCC3C4CCC([C@H](C)CCC(=O)OC)C4(C)[C@@H](O)CC3C1(C)CC[C@H](C2)OC(C)=O
InChI
1S/C27H44O5/c1-16(6-11-25(30)31-5)21-9-10-22-20-8-7-18-14-19(32-17(2)28)12-13-26(18,3)23(20)15-24(29)27(21,22)4/h16,18-24,29H,6-15H2,1-5H3/t16-,18?,19-,20?,21?,22?,23?,24+,26?,27?/m1/s1
InChI key
FVYCOKNQUSFTBH-HJPVDDEFSA-N
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C2525-500MG | 04061832269894 |