MilliporeSigma
All Photos(1)

C40300-HH

Sigma-Aldrich

DMT-dC(ac)-CPG

pore size 1000 Å, 25-35 μmol/g

type

for DNA synthesis

product line

Proligo Reagents

assay

≥95.0% (Nucleoside Purity, reversed phase HPLC)

form

solid

packaging

pkg of 1 × 1 g C403001-01
pkg of 1 × 10 g C403010-01

pore size

1000 Å

capacity

≥99.0 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service