MilliporeSigma
All Photos(2)
Key Documents
C40
Sigma-Aldrich
Catalase from bovine liver
lyophilized powder, ≥10,000 units/mg protein
Synonym(s):
H2O2:H2O2 oxidoreductase
Sign Into View Organizational & Contract Pricing
Select a Size
100 MG
$79.30
500 MG
$233.00
1 G
$401.00
$79.30
Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0041
Request a Bulk Order
Select a Size
Change View
100 MG
$79.30
500 MG
$233.00
1 G
$401.00
About This Item
CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
$79.30
Check Cart for Availability
Diagnostics Manufacturers should use alternative grade product: SRE0041
Request a Bulk Order
form
lyophilized powder
Quality Level
specific activity
≥10,000 units/mg protein
mol wt
tetramer ~250 kDa
contains
≤0.2 wt. % Thymol
isoelectric point
5.4
solubility
50 mM potassium phosphate buffer, pH 7.0: soluble 2.0 mg/mL
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
SMILES string
O(CC)C(=O)c1ccc(cc1)O
InChI
1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3
InChI key
NUVBSKCKDOMJSU-UHFFFAOYSA-N
Gene Information
cow ... CAT(280743)
Looking for similar products? Visit Product Comparison Guide
General description
Research Area: Cell Signaling
Catalase from bovine liver is a tetramer consisting of 4 equal subunits each with a 60 kDa molecular weight. Each of these subunits contains iron bound to a protoheme IX group. The enzyme will also strongly bind to NADP, where NADP and the heme group are within 13.7 angstroms.
Application
Catalase acts as a natural antioxidant to study the roles of reactive oxygen species in gene expression and apoptosis. It has also been used to protect against oxidative damage to proteins, lipids, and nucleic acids. Industrially, catalzes have been used to remove hydrogen peroxide added to milk and cheese, in textile bleaching, and to examine its positive effects on the viability of DNA-repair mutants of E. coli.
Catalase from bovine liver may be used:
  • to prepare H2O2-O2 based biocathode for applications in glucose biofuel cells
  • to study the kinetic properties and storage stability of catalase immobilized on to florisil
  • in glutathione-mediated superoxide generation in an aqueous solution
Biochem/physiol Actions
Catalase activity is constant over the pH range of 4.0-8.5. The enzyme activity is inhibited by 3-amino-1-H-1,2,4 triazole, cyanide, azide, hydroxylamine, cyanogen bromide, 2-mercaptoethanol, dithiothreitol, dianisidine, and nitrate. Incubation of catalase with ascorbate or ascorbate/Cu2+ results in degradation of the catalase molecule.
Catalase, an antioxidant enzyme found in all aerobic organisms, catalyzes the degradation of hydrogen peroxide, a byproduct of metabolic processes, into less harmful water and oxygen. It can also react with alkylhydrogen peroxides, such as methylperoxide and ethylperoxide and the second H2O2 molecule can be replaced by methanol, ethanol, propanol, formate and nitrate as a hydrogen donor. Catalase enzyme uses either iron (Fe) or manganese (Mn) as cofactor, and are classified as Fe-CAT or Mn-CAT.
Caution
Solutions of catalse should not be frozen. Frozen solution will result in a 50-70% loss of activity.
Unit Definition
One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 concentration falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.
Other Notes
The protein content is determined by the Biuret method. In the product specifications, a protein content >= 70% is guaranteed. The impurities are not analyzed. The only other content specification is the concentration of Thymol, <= 0.2% wt.
inhibitor
Product No.
Description
Pricing
pictograms
Health hazard
signalword
Danger
hcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Yifan Jiang et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-12)
Clematis florida is a popular ornamental vine species known for diverse colors and shapes of its flowers but not for scent. Here we investigated the composition and biosynthesis of floral scent in 'Kaiser', a fragrant cultivar of C. florida that
Emilia M Sogin et al.
mSystems, 4(6) (2019-12-12)
Microbial communities exchange molecules with their environment, which plays a major role in regulating global biogeochemical cycles and climate. While extracellular metabolites are commonly measured in terrestrial and limnic ecosystems, the presence of salt in marine habitats limits the nontargeted
Hanming Yang et al.
The Journal of chemical physics, 151(24), 245102-245102 (2020-01-03)
Low-concentration photochemically induced dynamic nuclear polarization (LC-photo-CIDNP) has recently emerged as a powerful technology for the detection of aromatic amino acids and proteins in solution in the low-micromolar to nanomolar concentration range. LC-photo-CIDNP is typically carried out in the presence
Saskia F Erttmann et al.
Nature communications, 10(1), 3493-3493 (2019-08-04)
Hydrogen peroxide (H2O2) has a major function in host-microbial interactions. Although most studies have focused on the endogenous H2O2 produced by immune cells to kill microbes, bacteria can also produce H2O2. How microbial H2O2 influences the dynamics of host-microbial interactions
Afra Khiralla et al.
Microorganisms, 8(9) (2020-09-10)
An endophytic fungus isolated from Vernonia amygdalina, a medicinal plant from Sudan, was taxonomically characterized as Curvularia papendorfii. Ethyl acetate crude extract of C. papendorfii revealed an important antiviral effect against two viral pathogens, the human coronavirus HCoV 229E and
Protocols
This procedure may be used for all Catalase products.
Questions
  1. Nelle specifiche leggo che il contenuto proteico di questa catalasi liofilizzata è del 70% Il restante 30% da cosa è composto ?
    1 answer
    1. The protein content is determined by the Biuret method. In the product specifications, a protein content >= 70% is guaranteed. As an example, lot number SLCL9662 reports a 90% protein content on the Certificate of Analysis (COA). The impurities are not analyzed. The only other content specification is the concentration of Thymol, <= 0.2% wt.
      Helpful?
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service