Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C4633

Sigma-Aldrich

Cholesteryl n-decanoate

~95% (HPLC; detection at 205 nm)

Synonym(s):

3β-Hydroxy-5-cholestene 3-decanoate, 5-Cholesten-3β-ol 3-decanoate, Cholesteryl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64O2
CAS Number:
Molecular Weight:
540.90
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

~95% (HPLC; detection at 205 nm)

form

solid

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCC(O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCCC(C)C)=O

InChI

1S/C37H64O2/c1-7-8-9-10-11-12-13-17-35(38)39-30-22-24-36(5)29(26-30)18-19-31-33-21-20-32(28(4)16-14-15-27(2)3)37(33,6)25-23-34(31)36/h18,27-28,30-34H,7-17,19-26H2,1-6H3

InChI key

LJGMGXXCKVFFIS-UHFFFAOYSA-N

Biochem/physiol Actions

Cholesteryl n-decanoate is an ester of cholesterol that interferes with the absorption of cholesterol. The level of cholesterol in liver and plasma in leghorn chicks was decreased when fed on diet of Cholesteryl n-decanoate.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Pattabhi et al.
Journal of lipid research, 20(6), 753-759 (1979-08-01)
Cholesteryl decanoate (C37H64O2) is monoclinic, space group P2I, with cell dimensions a = 12.931 (6), b = 9.066 (2), c = 30.22 (1) A, beta = 91.14 (4) degrees, and Z = 4. The atomic coordinates from cholesteryl laurate were
M P McCourt et al.
Journal of lipid research, 35(4), 584-591 (1994-04-01)
This paper describes the X-ray crystal structure analysis of a cholesteryl ester solid solution, cholesteryl decanoate/cholesteryl laurate, grown from a bulk concentration with molar ratio 0.56/0.43. The unit cell is monoclinic with a = 12.969, b = 9.048, c =
Dietary constituents affecting plasma and liver cholesterol in cholesterol-fed chicks.
D W PETERSON et al.
The Journal of nutrition, 50(2), 191-201 (1953-06-01)
Akio Sugihara et al.
Bioscience, biotechnology, and biochemistry, 66(11), 2347-2355 (2003-01-01)
With the aim of developing a new cholesterol esterase for eliminating lipids on used contact lenses, microorganisms were screened for the enzyme activity. A Pseudomonas aeruginosa isolated from soil was found to produce a desirable enzyme. The enzyme had an
W Guo et al.
Biochemistry, 32(35), 9038-9052 (1993-09-07)
Cholesteryl esters are a major lipid constituent of plasma lipoproteins and atherosclerotic lesions. Crystalline and liquid crystalline phases of several cholesteryl esters [oleate (C18:1, omega-9), erucate (C22:1, omega-9), hexanoate (C6:0), decanoate (C10:0), undecanoate (C11:0), myristate (C14:0), palmitate (C16:0), and stearate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service