MilliporeSigma
All Photos(1)

Documents

BICR 16

06031001

Sign Into View Organizational & Contract Pricing

Synonym(s):
BICR-16 R Cells, BICR16 Cells, Beatson Institute for Cancer Research 16 Cells

biological source

human mouth

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial, keratinocyte

products

Not specified

receptors

Epidermal Growth Factor Receptor (EGFR)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Tongue squamous carcinoma

Cell Line Description

Adherent cell line derived from a recurrent squamous cell carcinoma of the tongue of a Caucasian male. Keratin and involucrin markers present. Known mutations: p53, p16 and p14ARF. Cells are tumourigenic in athymic mice.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 0.4 micrograms/ml Hydrocortisone.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 using 0.05% trypsin/EDTA; 8% CO2; 37°C. Suggested seeding density 2-4 x 10,000 cells/cm2. A feeder layer of lethally-irradiated or mitomycin C-treated 3T3 feeder cells can be used to culture this cell line, howev

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service