MilliporeSigma
All Photos(1)

Documents

U2A cell line human

12021502

Sign Into View Organizational & Contract Pricing

Select a Size

biological source

human fibrosarcoma

growth mode

Adherent

morphology

Epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For complete product information, please see ECACC

Cell Line Description

The human sarcoma cell line HT 1080 (ECACC Catalogue number 85111505) was transfected with a vector encoding a selectable marker (guanine phosphoribosyltransferase) regulated by alpha interferon to create the 2fTGH cell line, enabling the selection of mutations in genes encoding components of the interferon signalling pathway. Chemical mutagenesis of the parent 2fTGH cell line enabled the isolation of a number of IFNg signalling mutants available from ECACC: U2A (12021502), U3A (12021503), U4A (12021504), U4C (12021505), U5A (12021506), U6A (12021507). Each contains a different mutation in the IFN signalling pathway. The 2fTGH panel provides a tool for the in vitro study and comparison of disrupted interferon signalling at multiple points across the IFN pathway. The U2A mutant cell line lacks expression of p48 gene product. U2A is unresponsive to interferon-alpha and interferon-beta; furthermore it is partially defective in response to interferon-gamma.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 1-4x10,000 cells/cm² using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service