Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C16 (HeLa derivative)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human cervix

packaging

tube of 5 μg 84121902-DNA-5UG
pkg of vial of cells 84121902-1VL

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

This was originally a clone of the MRC-5 cell line. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.

Application

Transformation studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service