CLS430720

Corning® cell culture flasks

surface area 75 cm2, canted neck, cap (plug seal)

Synonym(s):
flask cell culture, tissue culture flask, culture flask
eCl@ss:
32040501

material

canted neck
cap (plug seal)
polystyrene
rectangular flask

sterility

sterile

feature

graduations

packaging

case of 100

manufacturer/tradename

Corning 430720

maximum volume

60 mL

surface area

75 cm2

working volume

15-22.5 mL

matrix active group

TC-Treated surface treatment

Looking for similar products? Visit Product Comparison Guide

General description

Available in a variety of sizes, designs and cap styles to meet your needs.
  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Printed with lot numbers for ease in traceability
  • 100% integrity tested
  • Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service