MilliporeSigma
All Photos(2)

Documents

CN352-05

Sigma-Aldrich

Canine Pulmonary Artery Smooth Muscle Cells: CnPASMC (Cryovial)

Sign Into View Organizational & Contract Pricing

NACRES:
NA.81

biological source

canine pulmonary artery (normal, tunica intima and media)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 78

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P352-05CN350-05CN150-05
packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells

growth mode

Adherent

growth mode

Adherent

growth mode

-

growth mode

Adherent

karyotype

2n = 78

karyotype

2n = 38

karyotype

2n = 78

karyotype

2n = 78

morphology

smooth muscle

morphology

smooth muscle

morphology

-

morphology

Skeletal muscle

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Canine Pulmonary Artery Smooth Muscle Cells (CnPASMC) provide a solid animal model to examine physiology of vascular smooth muscle cells, as well as the function of pulmonary arteries in both normal and disease conditions.

Cell Line Origin

Artery

Application

cell growth, signaling, receptor transactivation and suppression, vascular remodeling

Components

Canine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Canine Smooth Muscle Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the CnPASMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service