MilliporeSigma
All Photos(7)

Documents

CWA-1160

Sigma-Aldrich

Anti-Mouse OX40L/CD252 (RM134L) ColorWheel® Dye-Ready mAb

greener alternative
Sign Into View Organizational & Contract Pricing

Synonym(s):
Tumor necrosis factor ligand superfamily member 4;OX40 Ligand

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM134L, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 22.26 kDa

purified by

affinity chromatography

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Ox40l(22164)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CWA-1170CWA-1136CWA-1168
species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

species reactivity

human

biological source

rat

biological source

rat

biological source

rat

biological source

mouse

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

technique(s)

flow cytometry: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Still not finding the right product?  

Give our Product Selector Tool a try.

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone RM134L is a rat monoclonal antibody that detects OX40L/CD252.

Immunogen

Rat NRK-52E cells transfected with mouse OX-40L/CD252.

Application

Quality Control Testing

Evaluated by Flow Cytometry in CHOZN cells stably transfected with mouse OX40L/CD252.

Flow Cytometry Analysis: Staining of one million CHOZN cells stably transfected with mouse OX40L/CD252 was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1160, Anti-Mouse OX40L/CD252 (RM134L) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1012 Rat IgG2b isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Tested Applications

Flow Cytometry Analysis: Staining of one million CHOZN cells stably transfected with mouse OX40L/CD252 was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1160, Anti-Mouse OX40L/CD252 (RM134L) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1012 Rat IgG2b isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Flow Cytometry Analysis: Staining of one million L5178Y cells stably transfected with mouse OX40L/CD252 was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1160, Anti-Mouse OX40L/CD252 (RM134L) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1012 Rat IgG2b isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Tumor necrosis factor ligand superfamily member 4 (UniProt: P43488; also known as OX40 Ligand, OX40L, CD252) is encoded by the Tnfsf4 (also known as Ox40l, Txgp1l) gene (Gene ID: 22164) in murine species. OX40L, a member of the tumor necrosis factor family, is a single-pass type II membrane protein that contains a cytoplasmic domain (aa 1-28), a transmembrane domain (aa 29-50), and an extracellular domain (aa 51-198). It is a co-stimulatory molecule that binds to OX40 (TNFDF4) expressed on T cells, NK cells, B cells, and NKT cells. OX40L is broadly expressed by antigen presenting cells, activated T cells, and endothelial cells. OX40-OX40L interaction plays a key role in inducing T helper cells 2 (Th2) polarization and inhibiting T regulatory cell development. This interaction is also involved in many aspects of physiological responses between T cells and lymphoid and non-lymphoid cells. OX40/OX40L interactions is reported to play a key role in the development of multiple inflammatory and autoimmune diseases, making them attractive candidates for drug development. The absence of OX40/OX40L interactions is reported to suppress immune disease development in animal models. This interaction also plays a crucial role in the pathogenesis of experimental allergic encephalomyelitis (EAE). Mice deficient in OX40L exhibit poorly developed EAE, and OX40L transgenic mice display enhanced EAE symptoms. Blockade of OX40-OX40L interaction in arthritic mice is shown to reduce inflammation and restore tissue integrity predominantly by inhibiting inflammatory cytokine production by OX40L-expressing macrophages. (Ref.: Findlay, EG., et al. (2014). Proc. Natl. Acad. Sci. USA. 111(6); 2289-2294; Croft, M., et al. (2009). Immunol. Rev. 229(1); 173-191; Baum, PR., et al. (1994). EMBO J. 13(17); 3992-4001).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 µL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 µL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Protocols

ColorWheel® protocol guides flow cytometry analysis with ColorWheel® antibodies and dyes for precise results.

Related Content

Unlock freedom in your flow cytometry assay design with ColorWheel® flow cytometry antibodies and dyes. The ColorWheel® flow cytometry portfolio was designed with simplicity and flexibility in mind to help streamline your flow cytometry workflow without compromising on quality.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service