MilliporeSigma
All Photos(1)

D042

Sigma-Aldrich

R(−)-N-Allylnorapomorphine hydrobromide

solid

Empirical Formula (Hill Notation):
C19H19NO2 · HBr
CAS Number:
Molecular Weight:
374.27
MDL number:
PubChem Substance ID:

form

solid

optical activity

[α]25/D −46.8°, c = 0.5 in H2O(lit.)

color

tan

solubility

H2O: 0.29 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.6 mg/mL (Solutions should be freshly prepared.)
0.1 M HCl: soluble (Solutions should be freshly prepared.)
ethanol: slightly soluble (Solutions should be freshly prepared.)

storage temp.

2-8°C

SMILES string

Br.Oc1ccc2C[C@H]3N(CCc4cccc(c34)-c2c1O)CC=C

Biochem/physiol Actions

Dopamine receptor agonist; intermediate for the preparation of tritium or deuterium labeled R(−)-N-propylnorapomorphine.

Caution

Photosensitive. Reseal vial under argon or nitrogen to maximize stability.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service