Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCC(=O)CC[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O
Biochem/physiol Actions
Enzyme derived prostaglandin that acts as a hormone in uterine functions and may effect glucose uptake.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Huifen Wang et al.
Obesity (Silver Spring, Md.), 19(12), 2404-2410 (2011-07-23)
Pentadecanoic acid (15:0) and heptadecanoic acid (17:0), the dairy-specific saturated fatty acids have been inversely, while inflammation and oxidative stress have been positively related to the risk of cardiovascular disease (CVD). Both fatty acid metabolism and inflammation and oxidative stress
H T Duong et al.
Reproduction in domestic animals = Zuchthygiene, 47(6), 939-945 (2012-02-18)
Previous in vitro studies demonstrated that bovine endometrium has the capacity to convert inactive cortisone to biologically active cortisol (Cr) and that Cr inhibits cytokine-stimulated prostaglandin F(2α) (PGF) production. This study was carried out to test the hypothesis that bovine
Hemanta K Shrestha et al.
Biology of reproduction, 85(3), 482-489 (2011-06-10)
A bolus treatment (e.g., 25 mg) of prostaglandin F(2alpha) (PGF) in the study of luteolysis in cattle results in dubious interpretations. Therefore, in experiment 1 of the present study, a 13,14-dihydro-15-keto-PGF (PGFM) pulse was simulated by incremental intrauterine (IU) infusion
G Pugliesi et al.
Theriogenology, 76(4), 640-651 (2011-05-27)
Flunixin meglumine (FM; 2.5 mg/kg) was given to heifers at three 8-h intervals, 16 d after ovulation (first treatment = Hour 0) to inhibit the synthesis of prostaglandin F(2α) (PGF), based on plasma concentrations of a PGF metabolite (PGFM). Blood
G Pugliesi et al.
Domestic animal endocrinology, 41(2), 98-109 (2011-07-12)
Various doses of estradiol-17β (E(2)) were used in heifers to induce a pulse of 13,14-dihydro-15-keto-prostaglandin F(2α) (PGFM). The effect of E(2) concentration on the prominence of PGFM pulses and the relationship between prominence and intrapulse concentration of progesterone (P(4)), LH
Related Content
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service