Skip to Content
MilliporeSigma
All Photos(1)

Documents

D8506

Sigma-Aldrich

Dansyl-L-cysteic acid

Sign Into View Organizational & Contract Pricing

Empirical Formula (Hill Notation):
C15H18N2O7S2
CAS Number:
Molecular Weight:
402.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NC(CS(O)(=O)=O)C(O)=O

InChI

1S/C15H18N2O7S2/c1-17(2)13-7-3-6-11-10(13)5-4-8-14(11)26(23,24)16-12(15(18)19)9-25(20,21)22/h3-8,12,16H,9H2,1-2H3,(H,18,19)(H,20,21,22)

InChI key

ZVOGQJUKPIENKH-UHFFFAOYSA-N

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service