E7627

Sigma-Aldrich

Ethyl erucate

~99% (capillary GC), liquid

Synonym(s):
cis-13-Docosenoic acid ethyl ester
Empirical Formula (Hill Notation):
C24H46O2
CAS Number:
Molecular Weight:
366.62
EC Number:
MDL number:
PubChem Substance ID:

assay

~99% (capillary GC)

form

liquid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC

InChI

1S/C24H46O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-4-2/h11-12H,3-10,13-23H2,1-2H3/b12-11-

InChI key

WFZQLUSOXHIVKL-QXMHVHEDSA-N

Analysis Note

Sealed ampule Isomer content given on label.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.