E7888

Sigma-Aldrich

N-Acetyl-eglin C from leeches

recombinant, expressed in proprietary host

Synonym(s):
Eglin c
Empirical Formula (Hill Notation):
C377H552N96O107
CAS Number:
Molecular Weight:
8141.00
MDL number:
Pricing and availability is not currently available.

recombinant

expressed in proprietary host

storage temp.

−20°C

General description

Structural dynamics have been intensively studied.

Biochem/physiol Actions

Inhibitor of chymotrypsin, subtilisin, leukocyte elastase and cathepsin G.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
H Rink et al.
Nucleic acids research, 12(16), 6369-6387 (1984-08-24)
A DNA containing the coding sequence for the proteinase inhibitor protein, eglin c, from the leech Hirudo medicinalis has been obtained by enzymatic assembly of chemically synthesized DNA fragments. The synthetic gene consists of a 232 base-pair fragment containing initiation...
U Seemüller et al.
Hoppe-Seyler's Zeitschrift fur physiologische Chemie, 361(12), 1841-1846 (1980-12-01)
The leech Hirudo medicinalis contains three different groups of proteinase inhibitor proteins, the thrombin-specific hirudin, the bdellins directed against trypsin, plasmin and acrosin, and the eglins which were discovered only recently. We are interested in the eglins mainly for two...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.