Skip to Content
MilliporeSigma

EHU017931

MISSION® esiRNA

targeting human CTD-2600O9.1

Sign In to View Organizational & Contract Pricing.

Select a Size

20 μG

$242.00

50 μG

$435.00

$242.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

MISSION® esiRNA, targeting human CTD-2600O9.1

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACAAGACTCTGCCCATCACCCCCAACCTGGCTGATCTGGGACCACATCAGCACCTGGCCTGACTCTTCTACCAGGATGCCCTCCAAGACCAACACTGATGCCTTAGTCACCCTCACCAGGGAGGCTGGGGGTGCCCCCACATGGCTGCTGCACCTGATGTTGGAAACGGAGGCTCCAAAAACGTGAGAAAACCGAGGCCCGGAAAGTGTCAACCCAAATCACGACACTCCTTGCCCCAAACCCAGAAGCCTGAGGAAGTAAGGAAGTGCGGTGGGATCTGTCTTCCCGTGTGTCTTCAGGATCCTGGGGCCTGCCCCTCCTGGCTTCCTCTTGAGGAAGGCCTCAGAAGACCCGCCTTTGGTCACAGGACAGAAGCTGGGCCAAGAGGCAAAGGTTTTGGCCGCCACAGGGGAAACTCTGGGCTCAGAGCTGCCGAAGTGACTCACGGGCCCTGCTGGGAAGCAGCTGACGTTCCTTGTCTGCGCCTTACAAAACCACACGGGCT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Quality Level

Gene Information

human ... SERPINA1(5265)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU119921EHU071831EHU040611
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

ACAAGACTCTGCCCATCACCCCCAACCTGGCTGATCTGGGACCACATCAGCACCTGGCCTGACTCTTCTACCAGGATGCCCTCCAAGACCAACACTGATGCCTTAGTCACCCTCACCAGGGAGGCTGGGGGTGCCCCCACATGGCTGCTGCACCTGATGTTGGAAACGGAGGCTCCAAAAACGTGAGAAAACCGAGGCCCGGAAAGTGTCAACCCAAATCACGACACTCCTTGCCCCAAACCCAGAAGCCTGAGGAAGTAAGGAAGTGCGGTGGGATCTGTCTTCCCGTGTGTCTTCAGGATCCTGGGGCCTGCCCCTCCTGGCTTCCTCTTGAGGAAGGCCTCAGAAGACCCGCCTTTGGTCACAGGACAGAAGCTGGGCCAAGAGGCAAAGGTTTTGGCCGCCACAGGGGAAACTCTGGGCTCAGAGCTGCCGAAGTGACTCACGGGCCCTGCTGGGAAGCAGCTGACGTTCCTTGTCTGCGCCTTACAAAACCACACGGGCT

esiRNA cDNA target sequence

TAAAGCCTCGGCAGCACTATTTGGACAGCATCAAGGAGCTTTATGGAGCTTTTGCTTTTTCTGCCAACTTCACAGATTCTGTTACAACTGGGAGGCAGATTAATGACTATTTGAGAAGGCAAACATACGGGCAAGTCGTGGACTGCCTCCCGGAGTTCAGCCAGGACACGTTCATGGTTCTTGCCAATTACATCTTCTTCAAAGCCAAGTGGAAGCACCCTTTCAGTCGCTACCAGACCCAGAAGCAGGAAAGTTTCTTTGTGGATGAGAGGACTTCTCTCCAGGTCCCCATGATGCACCAAAAGGAAATGCACAGATTCCTCTATGACCAGGATTTGGCTTGCACCGTCCTCCAGATAGAATACAGAGGAAATGCCTTGGCGCTGCTGGTCCTCCCTGACCCGGGGAAAATGAAGCAGGTGGAGGCTGCTCTGCAGCCACAGACCCTGAGAAAATGGGGCCAATTGCTCCTGCCCAGTCTGTTGGATTTGCACTTGCCAAGGTT

esiRNA cDNA target sequence

GCAGCTACCACCACCAAGTTCATAGTCCGATCGAAGGATGGCCCCTCTTACTTCACTGTCTCCTTCAATAGGACCTTCCTGATGATGATTACAAATAAAGCCACAGACGGTATTCTCTTTCTAGGGAAAGTGGAAAATCCCACTAAATCCTAGGTGGGAAATGGCCTGTTAACTGATGGCACATTGCTAATGCACAAGAAATAACAAACCACATCCCTCTTTCTGTTCTGAGGGTGCATTTGACCCCAGTGGAGCTGGATTCGCTGGCAGGGATGCCACTTCCAAGGCTCAATCACCAAACCATCAACAGGGACCCCAGTCACAAGCCAACACCCATTAACCCCAGTCAGTGCCCTTTTCCACAAATTCTCCCAGGTAACTAGCTTCATGGGATGTTGCTGGGTTACCAT

esiRNA cDNA target sequence

GGTGGTCCTCATGGAGAAAATGGGTGACCACCTCGCCCTTGAAGACTACCTGACCACAGACTTGGTGGAGACATGGCTCAGAAACATGAAAACCAGAAACATGGAAGTTTTCTTTCCGAAGTTCAAGCTAGATCAGAAGTATGAGATGCATGAGCTGCTTAGGCAGATGGGAATCAGAAGAATCTTCTCACCCTTTGCTGACCTTAGTGAACTCTCAGCTACTGGAAGAAATCTCCAAGTATCCAGGGTTTTACAAAGAACAGTGATTGAAGTTGATGAAAGGGGCACTGAGGCAGTGGCAGGAATCTTGTCAGAAATTACTGCTTATTCCATGCCTCCTGTCATCAAAGTGGACCGGCCATTTCATTTCATGATCTATGAAGAAACCTCTGGAATGCTTCTGTTTCTGGGCAGGGTGGTGAATCCGACTCTCCTATAATTCAGGACACGCATAAGCACTTCGTG

Ensembl | human accession no.

ENSG00000187185

Ensembl | human accession no.

ENSG00000186910

Ensembl | human accession no.

ENSG00000170054

Ensembl | human accession no.

ENSG00000140093

Gene Information

human ... SERPINA1(5265)

Gene Information

human ... SERPINA11(256394), SERPINA11(256394)

Gene Information

human ... SERPINA9(327657), SERPINA9(327657)

Gene Information

human ... SERPINA10(51156), SERPINA10(51156)

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Instructions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service