Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GTGGCTCCACCCACAAGGTTAAGATATAATGTAATATCTCATGACAGTATACAGATTTCATGGAAGGCTCCAAGAGGGAAATTTGGTGGTTACAAACTTCTTGTGACTCCAACTTCAGGTGGAAAAACTAACCAGCTGAATCTGCAGAACACTGCAACTAAAGCAATTATTCAAGGCCTTATGCCAGACCAGAATTACACAGTTCAAATTATTGCATACAATAAAGATAAAGAAAGCAAGCCAGCTCAAGGCCAATTCAGAATTAAAGATTTAGAAAAAAGAAAGGATCCAAAGCCCAGAGTCAAAGTTGTGGACAGAGGAAATGGGAGTAGACCATCTTCACCAGAAGAAGTGAAATTTGTCTGTCAAACTCCAGCAATTGCTGACATTGTAATCCTGG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Quality Level
Related Categories
1 of 4
This Item | EHU097091 | EHU096651 | EHU093321 |
|---|---|---|---|
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence GTGGCTCCACCCACAAGGTTAAGATATAATGTAATATCTCATGACAGTATACAGATTTCATGGAAGGCTCCAAGAGGGAAATTTGGTGGTTACAAACTTCTTGTGACTCCAACTTCAGGTGGAAAAACTAACCAGCTGAATCTGCAGAACACTGCAACTAAAGCAATTATTCAAGGCCTTATGCCAGACCAGAATTACACAGTTCAAATTATTGCATACAATAAAGATAAAGAAAGCAAGCCAGCTCAAGGCCAATTCAGAATTAAAGATTTAGAAAAAAGAAAGGATCCAAAGCCCAGAGTCAAAGTTGTGGACAGAGGAAATGGGAGTAGACCATCTTCACCAGAAGAAGTGAAATTTGTCTGTCAAACTCCAGCAATTGCTGACATTGTAATCCTGG | esiRNA cDNA target sequence GATTTCATGGAAGGCTCCAAGAGGGAAATTTGGTGGTTACAAACTTCTTGTGACTCCAACTTCAGGTGGAAAAACTAACCAGCTGAATCTGCAGAACACTGCAACTAAAGCAATTATTCAAGGCCTTATGCCAGACCAGAATTACACAGTTCAAATTATTGCATACAATAAAGATAAAGAAAGCAAGCCAGCTCAAGGCCAATTCAGAATTAAAGATTTAGAAAAAAGAAAGGATCCAAAGCCCAGAGTCAAAGTTGTGGACAGAGGAAATGGGAGTAGACCATCTTCACCAGAAGAAGTGAAATTTGTCTGTCAAACTCCAGCAATTGC | esiRNA cDNA target sequence ATCCTGGAGCATTGGAGATGAATTTCAATAAGATCATCAGCTTTCTATACAGCACTGTTGGAGCCCTGAACAAGATTGGCACAGATGGAACCCAAGTTGCAATGGTTCAGTTCACTGATGATCCCAGAACAGAATTTAAACTAAATGCTTACAAAACCAAAGAGACTCTTCTTGATGCAATTAAACACATTTCATACAAAGGAGGAAATACAAAAACAGGAAAAGCAATTAAGTATGTTCGAGATACCTTGTTCACTGCAGAGTCAGGTACAAGAAGGGGCATCCCAAAGGTTATCGTGG | esiRNA cDNA target sequence GAGCTGTTGCTGTGTTCTGCAGAGCTGAGCGAGTTCACCACAGTGGTGGGGAACACCATGTGCACCTTGGACTTCTATGAAGGGCAAGGCCGTCTGGATGGGGCTCTCTGCTCCTACACGGAGAAGGACAAGCAGGCGTATCTGGAGGCAGCCTATGCAGCCGGCGTCCGCAATATCGAGATGGAGTCCTCGGTGTTTGCCGCCATGTGCAGCGCCTGCGGCCTCCAAGCGGCCGTGGTGTGTGTCACCCTCCTGAACCGCCTGGAAGGGGACCAGATCAGCAGCCCTCGCAATGTGCTCAGCGAGTACCAGCAGAGGCCGCAGCGGCTGGTGAGCTACTTCATCAAGAAGAAACTGAGCAAGGCCTGAG |
| Gene Information human ... COL14A1(7373), COL14A1(7373) | Gene Information human ... COL14A1(7373), COL14A1(7373) | Gene Information human ... COL14A1(7373), COL14A1(7373) | Gene Information human ... UPP1(7378), UPP1(7378) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service