Skip to Content
MilliporeSigma

EHU148921

MISSION® esiRNA

targeting human IGLL1

Sign In to View Organizational & Contract Pricing.

Select a Size

20 μG

$242.00

50 μG

$435.00

$242.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGTTTTAAGTCAGCCCAAGGCCACCCCCTCGGTCACTCTGTTCCCGCCGTCCTCTGAGGAGCTCCAAGCCAACAAGGCTACACTGGTGTGTCTCATGAATGACTTTTATCCGGGAATCTTGACGGTGACCTGGAAGGCAGATGGTACCCCCATCACCCAGGGCGTGGAGATGACCACGCCCTCCAAACAGAGCAACAACAAGTACGCGGCCAGCAGCTACCTGAGCCTGACGCCCGAGCAGTGGAGGTCCCGCAGAAGCTACAGCTGCCAGGTCATGCACGAAGGGAGCACCGTGGAGAAGACGGTGGCCCCTGCAGAATGTTCATAGGTTCCCAGCCCCGACCCCACCCAAAGGGGCCTGGAGCTGCAGGATCCCAGGGGAAGGGTCTCTCTCTGCATCCCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Quality Level

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU077151EHU106581EHU034911
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | human accession no.

ENSG00000128322

Ensembl | human accession no.

ENSG00000147255

Ensembl | human accession no.

ENSG00000120235

Ensembl | human accession no.

ENSG00000103375

Gene Information

human ... IGLL1(3543), IGLL1(3543)

Gene Information

human ... IGSF1(3547), IGSF1(3547)

Gene Information

human ... IFNA6(3443), IFNA6(3443)

Gene Information

human ... AQP8(343), AQP8(343)

esiRNA cDNA target sequence

CCGTTTTAAGTCAGCCCAAGGCCACCCCCTCGGTCACTCTGTTCCCGCCGTCCTCTGAGGAGCTCCAAGCCAACAAGGCTACACTGGTGTGTCTCATGAATGACTTTTATCCGGGAATCTTGACGGTGACCTGGAAGGCAGATGGTACCCCCATCACCCAGGGCGTGGAGATGACCACGCCCTCCAAACAGAGCAACAACAAGTACGCGGCCAGCAGCTACCTGAGCCTGACGCCCGAGCAGTGGAGGTCCCGCAGAAGCTACAGCTGCCAGGTCATGCACGAAGGGAGCACCGTGGAGAAGACGGTGGCCCCTGCAGAATGTTCATAGGTTCCCAGCCCCGACCCCACCCAAAGGGGCCTGGAGCTGCAGGATCCCAGGGGAAGGGTCTCTCTCTGCATCCCAA

esiRNA cDNA target sequence

AGTGATGGACCCTCAACCAGAGTTGTGGATAGAGTCCAACTACCCCCAGGCCCCTTGGGAGAACATCACGCTTTGGTGCCGAAGCCCCTCTCGGATATCAAGCAAGTTCCTGCTGCTGAAGGATAAGACACAGATGACCTGGATCCGCCCTTCCCACAAGACCTTCCAAGTTTCATTCCTTATAGGTGCCCTTACTGAGTCCAATGCAGGTCTTTACCGGTGCTGCTACTGGAAGGAGACAGGCTGGTCAAAGCCCAGTAAAGTTCTAGAGTTGGAGGCACCAGGCCAACTGCCCAAGCCCATCTTCTGGATTCAGGCTGAGACCCCCGCTCTTCCTGGGTGTAATGTTAACATCCTCTGCCATGGCTGGCTGCAGGATTTGGTATTCATGCTGTTTAAAGAGGGATATGCAGAGCCTGTGGATTACCAAGTCCCAACTGGGA

esiRNA cDNA target sequence

CTGGACTGTGATCTGCCTCAGACCCACAGCCTGGGTCACAGGAGGACCATGATGCTCCTGGCACAAATGAGGAGAATCTCTCTTTTCTCCTGTCTGAAGGACAGACATGACTTCAGATTTCCCCAGGAGGAGTTTGATGGCAACCAGTTCCAGAAGGCTGAAGCCATCTCTGTCCTCCATGAGGTGATTCAGCAGACCTTCAACCTCTTCAGCACAAAGGACTCATCTGTTGCTTGGGATGAGAGGCTTCTAGACAAACTCTATACTGAACTTTACCAGCAGCTGAATGACCTGGAAGCCTGTGTGATGCAGGAGGTGTGGGTGGGAGGGACTCCCCTGATGAATGAGGACTCCATCCTGGCTGTGAGAAAATACTTCCAAAGAATCACTCTCTACCTGACAGAGAAAAAGTACAGCCCTTGTGCCTG

esiRNA cDNA target sequence

TCATTGGAGATGGGAAGACCCGCCTCATCCTGAAGGCTCGGTGAAGCAGAGCTCGTGGGATTCCTGCTGCTCCAGGTGTCCTCAGCTCACCTGTCCCAGACTGAGGACAGGGGAGTTCCTGCATTTCCTGCCAGGGCAGAGGCCCAGAGGAGCGACCCCCTGCTTCCACTGCTTGGGCCTGCTTTCTCAGATAGACTGACTGCTGAGGAGGCTCTAGGTTCTTGGAATTCCTTTGTGCTCATCAGAGACCCCAGCCTGGGGAACACGCTGCCCGCACTGCCCAGAGAGCAGTGCAAACACCACAACACGAGCGTGTTTCTTGAGAGGAATGTCCCCGAGTTGGACAAGGAGGCTGTTTCTGCACATCAGCTCATTTCCCGCACCCCATTTCTTGCTTGATTGCTTTGTTGG

NCBI accession no.

NM_020070

NCBI accession no.

NM_001555

NCBI accession no.

NM_021002

NCBI accession no.

NM_001169

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service