Skip to Content
MilliporeSigma

EMU037571

MISSION® esiRNA

targeting mouse Aqp9

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAGTCGAGAAAAGGCTGGTGGGATCATCACTATCAATATTGGATTTGCAACGGCAGTTGTGATGGCTCTTTATGCGACTTTTGGTGTCTCTGGGGGCCACATCAACCCAGCGGTGTCTTTTGCAATGTGCACCTTTGGAAGGATGGAGTGGTTCAAGTTCCCCTTTTATGTGGGAGCTCAGCTTTTGGGAGCCTTTGTCGGGGCTGCAACTGTCTTTGGCATTTACTATGACGGACTCATGGCCTTTGCTGATGGAAAGCTGCTCATCACGGGAGAAAATGGAACGGCGTTCATTTTCGCGACATACCCAAAACCATTTGTATCCGTGCCAGGTGCCTTTGTAGACCAAGTGGTGTCTACCATGTTCCTCCTTCTGATTGTCTTTGCCATCTTTGACTCCAGAAACCTGGGGGTCCCCAGAGGCCTGGAGCCCATTGTCATTGGCCTCCTAATCATTGTCATTTCCTGTTCTCTCGGACTCAACTCTGGTTGTGCCA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU016941EMU090951EMU037751
Ensembl | mouse accession no.

ENSMUSG00000032204

Ensembl | mouse accession no.

ENSMUSG00000096054

Ensembl | mouse accession no.

ENSMUSG00000075054

Ensembl | mouse accession no.

ENSMUSG00000004655

esiRNA cDNA target sequence

TCAGTCGAGAAAAGGCTGGTGGGATCATCACTATCAATATTGGATTTGCAACGGCAGTTGTGATGGCTCTTTATGCGACTTTTGGTGTCTCTGGGGGCCACATCAACCCAGCGGTGTCTTTTGCAATGTGCACCTTTGGAAGGATGGAGTGGTTCAAGTTCCCCTTTTATGTGGGAGCTCAGCTTTTGGGAGCCTTTGTCGGGGCTGCAACTGTCTTTGGCATTTACTATGACGGACTCATGGCCTTTGCTGATGGAAAGCTGCTCATCACGGGAGAAAATGGAACGGCGTTCATTTTCGCGACATACCCAAAACCATTTGTATCCGTGCCAGGTGCCTTTGTAGACCAAGTGGTGTCTACCATGTTCCTCCTTCTGATTGTCTTTGCCATCTTTGACTCCAGAAACCTGGGGGTCCCCAGAGGCCTGGAGCCCATTGTCATTGGCCTCCTAATCATTGTCATTTCCTGTTCTCTCGGACTCAACTCTGGTTGTGCCA

esiRNA cDNA target sequence

CGGGAATTTCCTGATTGAAACCTGTGATGAGATTGTTTCTCGAGACCTTAAACAGCAGTTGCTCTTATTGAACGGTCGATGGAGGGAGCTCTTCATGGAAGTCAAGCAATATGCAAGAGCAGACGAGATGGACAGAATGAAGAAAGAGTATATAGACGTCACTACCACCCTCTTTGGTTTTGCAACTGAAGCCCACAGGAAACTCTCTGAGCCTTTAGAAGTCTCTTTTATCAATGTCAAGTTATTAATTCAAGACTTGGAGGATCTGGAGAAGAGGGTGCCTGTGATGGATGCTCAGTACAAGATGATTGCCAAGAAGGCACATCTCTTTGCCAAAGAAAGTCCCCAAGAAGAAGCGAATGAAATGCTTACAACCATGTCCAAGCTCAAAGAACAGCTCTCCAAGGTCAAAGAATGTTGCTCCCCACTGTT

esiRNA cDNA target sequence

CAGCCACGAGTTACGGATTTATTGGACGCCATTGAGGATATGGACCTTTGTCACGTAGTTCCGTCAGACCAAGCGAAAGGTGACAGTAACGCTGAGATTAACACCGATTCTGACAGTCCAAATGGGATGGGTTGTTCACATCCAGAACATTGTAGACCAAAAGAGCATGCACATTTGGAAAAACCAAGCCTCTCTTGGATTCTGGAGCAGACATCCAGTTTAGTGAAACAGCTGGGAGTACCGGCAGACATATTACAGCACCTGAGACAACTATAAAATTACCTCCCCATTTCTAGTGGAAGTAATGCTCAGTTATTACTTAGAGCATTGTGTTCTGGATAAGGTAACGCTGATTGAACGAACACTTCCCCTGTAGTTACCCTTTGTGGACGGAATTGTT

esiRNA cDNA target sequence

CATGAAGGTGTGGACCAGTGGCCAGGTGGAGGAGTATGACCTGGATGCTGACGACATCAACTCCAGGGTGGAGATGAAGCCCAAATAGAGGAGGCTTGGCCTGGGCATCTATATGGGGGTGGGGCAGGGATGGGCGGAGGGAGGGGAGGGCCGAAATCATATTGTAGACACTCTGACAAGCTGACCAAAGTCACTCCCCAAGACCTGCCTGACCCACATGGTCTAGCCTTGTCTGGGGCAGTAATATCACTGTCTTTCTTTCTGTTTCCTGGTCTCAGAGCTTCCTGGGGACCAAGATTTATCAACTCAGCACCCCACTCCTTTGACATCATGGAGGAGTGAAAGTGAGGGACCCACCTGCTAGTTGTCCCCTCAGAGTGTGCTAGGAAGTCCCCCTCATTCCAAAGCTGCCCAGCAAGTCCCCCACCTTAGGTGCCTGGGATTCTGCCAATGTCGCTTTGTGCCTTTGACCGCAGCCTTCCTTGGGGTAGGCACCTTAT

Gene Information

mouse ... AQP9(64008), Aqp9(64008)

Gene Information

mouse ... SYNE1(64009), Syne1(64009)

Gene Information

mouse ... YAE1D1(67008), 1600012F09Rik(67008)

Gene Information

mouse ... AQP1(11826), Aqp1(11826)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service