Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
Product Name
MISSION® esiRNA, targeting mouse Map4k4
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AGAAGTGGCCAAGGGAAAGTATACCCTCTGATCAGCCGAAGACGGTTCCAGCAAATGGATGTGCTCGAAGGCCTAAATGTCCTGGTGACAATATCTGGCAAAAAGGATAAGTTACGTGTCTACTATTTATCCTGGCTAAGAAATAAGATTCTTCATAATGATCCAGAGGTTGAGAAGAAGCAGGGGTGGACCACTGTGGGCGACTTGGAAGGGTGCGTGCACTATAAAGTCGTAAAATATGAAAGAATCAAGTTTCTGGTAATTGCTTTGAAGAGTTCTGTGGAAGTCTATGCATGGGCACCGAAGCCATATCACAAATTTATGGCCTTTAAGTCATTTGGAGAACTGCTACATAAGCCATTACTGGTGGATCTCACTGTGGAGGAAGGCCAGAGGTTGAAAGTGATCTATGGATCCTGTGCGGGATTCCATGCTGTTGATGTGGAT
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Quality Level
Gene Information
mouse ... MAP4K4(26921), Map4k4(26921)
1 of 4
This Item | EMU060031 | EMU086771 | EMU039911 |
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| esiRNA cDNA target sequence AGAAGTGGCCAAGGGAAAGTATACCCTCTGATCAGCCGAAGACGGTTCCAGCAAATGGATGTGCTCGAAGGCCTAAATGTCCTGGTGACAATATCTGGCAAAAAGGATAAGTTACGTGTCTACTATTTATCCTGGCTAAGAAATAAGATTCTTCATAATGATCCAGAGGTTGAGAAGAAGCAGGGGTGGACCACTGTGGGCGACTTGGAAGGGTGCGTGCACTATAAAGTCGTAAAATATGAAAGAATCAAGTTTCTGGTAATTGCTTTGAAGAGTTCTGTGGAAGTCTATGCATGGGCACCGAAGCCATATCACAAATTTATGGCCTTTAAGTCATTTGGAGAACTGCTACATAAGCCATTACTGGTGGATCTCACTGTGGAGGAAGGCCAGAGGTTGAAAGTGATCTATGGATCCTGTGCGGGATTCCATGCTGTTGATGTGGAT | esiRNA cDNA target sequence TCTTCAGAGAGGGCGACTGTTGGATCTGTATGGAGCTCATGTCTACCTCGTTCGATAAGTTTTACAAATATGTATATAGTGTGTTAGATGACGTTATTCCGGAAGAGATCTTAGGCAAAATCACTTTAGCAACTGTGAAAGCACTAAACCACTTAAAAGAAAACTTGAAAATTATTCACAGAGACATCAAACCTTCCAATATTCTTCTGGACAGAAGTGGAAATATAAAGCTCTGTGATTTCGGCATCAGTGGACAGCTTGTGGACTCTATTGCCAAGACAAGAGATGCTGGGTGTAGGCCGTATATGGCACCTGAAAGAATAGACCCAAGTGCATCAAGACAAGGGTATGATGTCCGCTCTGATGTCTGGAGTTTGGGGATCACATTGTACGAGTTGGCC | esiRNA cDNA target sequence CCGGTGGAGACAGAAGGTAATACCAAAGCTTGAACCCTGAGGCCCTGTTGGCCCCCTGGGCACTGGCCCAGCCCCGGCCTCCAGGAATGGCCCTTGTGGACCCCGACATTTTTAATAAGGACCCTCGAGAACACTATGATCTACTGCAACGACTGGGTGGTGGAACTTATGGAGAAGTATTCAAGGCTCGCGACAAGGTGTCTAAGGACCTGGTGGCTTTGAAGATGGTGAAGATGGAGCCTGATGATGACGTCGCCACCCTTCAGAAGGAGATTCTCATGCTGAAAACTTGCCGGCACGCCAACATCGTGGCTTACCATGGCAGTTACCTCTGGCTGCAGAAGCTGTGGATCTGCATGGAATTCTGTGGAGCCGGTTCTCTCCAGGACATCTACCAAGTGACTGGCTCTCTGTCCGAGCTCCAGATCAGCTATGTGTGCCGGGAAGTGCTTCAGGGACTGGCCTACCTGCATTCAGAGAAGAAGATACATCGAGACATCAAGGGAGCCAACATC | esiRNA cDNA target sequence CAGCAGGAAATCACCATCCTTCGTGAATGCCGCCACCCCAATGTCGTGGCCTACATCGGCAGCTACCTCAGGAATGACCGATTGTGGATCTGTATGGAGTTCTGTGGAGGAGGGTCACTGCAGGAGATTTACCACGCCACTGGGCCCCTGGAAGAACGGCAGATTGCCTATGTTTGCCGGGAGGCACTGAAGGGGCTCCACCACCTGCATTCTCAGGGGAAAATTCACCGAGACATTAAGGGTGCCAACCTTCTGCTCACTCTGCAGGGTGATGTCAAGCTGGCGGACTTTGGGGTGTCAGGCGAGCTGACAGCATCTGTGGCCAAAAGACGTTCTTTCATTGGCACTCCATACTGGATGGCACCAGAGGTTGCCGCTGTGGAACGCAAGGGTGGCTACAATGAGCTGTGTGACGTCTGGGCCCTGGGCATCACTGCCATTGAGTTGGGCGAGCTGCAACCGCCACTGTTCCACCTGCACCCCATGAGGGCCTTGATGCTCATGTCGAAAAGCA |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| Gene Information mouse ... MAP4K4(26921), Map4k4(26921) | Gene Information mouse ... MAP2K4(26398), Map2k4(26398) | Gene Information mouse ... MAP4K1(26411), Map4k1(26411) | Gene Information mouse ... MAP4K2(26412), Map4k2(26412) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service