Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
Product Name
MISSION® esiRNA, targeting mouse Col16a1
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AGGCTGCAGCTGATCTCTCTGGACCAGCATGTTGACATCCTGGGCTCCCGGCCTGTGGGTGCTCGGGCTCTGGGCCACCTTCAGCCATGGGACAAATATAGGTGAGCGGTGCCCAACTTCACAGCAAGAAGGACTCAAGCTAGAACACAGTAGTGACCCATCAACCAACGTGACTGGCTTCAACCTAATCCGCCGACTGAACCTCATGAAGACATCTGCCATCAAGAAGATCCGCAACCCCAAGGGGCCACTCATCTTGAGGCTGGGGGCTGCCCCAGTGACCCAACCAACTCGACGAGTGTTCCCTCGAGGCCTCCCAGAGGAGTTTGCCCTGGTCCTGACAGTGCTTCTGAAGAAACACACCTTCCGGAACACATGGTACTTGTTCCAAGTGACTGATGCAAATGGGTACCCACAGATCTCCTTGGAAGTCAACAGCCAGGAGCGGAGTCTAGAGCTCCGGGCACAGGGCCAAGATGGTGACTTTGTGTCCTGCATCTTCCCAGT
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Quality Level
Gene Information
mouse ... COL16A1(107581), Col16a1(107581)
1 of 4
This Item | EMU049031 | EMU010451 | EMU079871 |
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information mouse ... COL16A1(107581), Col16a1(107581) | Gene Information mouse ... COL17A1(12821), Col17a1(12821) | Gene Information mouse ... COL18A1(12822), Col18a1(12822) | Gene Information mouse ... COL10A1(12813), Col10a1(12813) |
| esiRNA cDNA target sequence AGGCTGCAGCTGATCTCTCTGGACCAGCATGTTGACATCCTGGGCTCCCGGCCTGTGGGTGCTCGGGCTCTGGGCCACCTTCAGCCATGGGACAAATATAGGTGAGCGGTGCCCAACTTCACAGCAAGAAGGACTCAAGCTAGAACACAGTAGTGACCCATCAACCAACGTGACTGGCTTCAACCTAATCCGCCGACTGAACCTCATGAAGACATCTGCCATCAAGAAGATCCGCAACCCCAAGGGGCCACTCATCTTGAGGCTGGGGGCTGCCCCAGTGACCCAACCAACTCGACGAGTGTTCCCTCGAGGCCTCCCAGAGGAGTTTGCCCTGGTCCTGACAGTGCTTCTGAAGAAACACACCTTCCGGAACACATGGTACTTGTTCCAAGTGACTGATGCAAATGGGTACCCACAGATCTCCTTGGAAGTCAACAGCCAGGAGCGGAGTCTAGAGCTCCGGGCACAGGGCCAAGATGGTGACTTTGTGTCCTGCATCTTCCCAGT | esiRNA cDNA target sequence TTGGCAGAGCCACCAAGTATCTGGGACCGGTTCTTGGGCGCCTATGTCCAAGGGTCCCCGTCCTCCTGAGACCAAATCACCCAGCGACACTTGTTTTAGTCTGAACCAAATATATACATACAGAAAATTCAAAGACACACAATCTGAAACAGCTCCATTGGTTGGGTGGGCCTTTCGGAATAGCTGCAATGTTTAATATGAGACTTGACCTCTCTGCTACATAGATCTGAGCTCCCCGGCTTCTTTAGCATTTAGCCCAGCCTCTGCCAGGCCTCGGAAGCCAGTTACATAACTGTTGGCTGGGGATCTTGGAGCTTTGCCTAAAAAGTGAGCCCAGAAAATGGAAAATGGGAATGGGGACCTTTGTAGATGACGTGTGATGGGTTATGTTACGAAGGCTGCTGTCAGCTCCTCCTCCTCTC | esiRNA cDNA target sequence CAGGCTCCTGGAACAGAAAGCTGCGAGCTGCCACAACAGCTACATCGTCCTGTGCATTGAGAATAGCTTCATGACCTCTTTCTCCAAATAGGGCCTCTGCCAGCTAGGGTGGCAGACAGAGGCCATGCAGAACTTTGACACAGCGCAGGGAGCATTCAGTCAGCACCCAGGGCTCTGGCTGGGATACAATCCTGTATAGTTCCCATTTTTATGTAATCCTCAAGAAATAAAAGGAAGCCAAAGAGTATATTTTTCTAAAACATTTACAACAGACTTCCTGCCCTTCCTCTCCCACTTGAACTGGCCAGTCTGTGTCACAGTCCCTTCTTCCTCCGTGCTTGGTAGAGCTTCCTGGCGAAGTCGGACCACAGCTAGGTCTCCACCGGAGGCGATGCCCTACAGTTGCTGCCTCAGACCT | esiRNA cDNA target sequence TCATGCCTGATGGCTTCATAAAGGCAGGCCAGAGGCCCAGGCTTTCTGGGATGCCGCTTGTCAGTGCTAACCACGGGGTAACAGGTATGCCCGTGTCTGCTTTTACTGTCATTCTCTCTAAAGCTTACCCAGCAGTAGGTGCCCCCATCCCATTTGATGAGATTCTGTACAATAGGCAGCAGCATTACGACCCAAGATCTGGTATCTTTACCTGTAAGATCCCAGGCATATACTATTTCTCCTACCACGTGCATGTGAAAGGGACTCACGTTTGGGTAGGCCTGTATAAGAACGGCACGCCTACGATGTACACGTATGATGAGTACAGCAAAGGCTACCTGGATCAGGCTTCAGGGAGTGCAATCATGGAGCTCACAGAAAATGACCAGGTATGGCTCCAATTGCCCAATGCAGAATCAAACGGCCTCTACTCCTCTGAGTACGTCCACTCGTCCTTCTCAGG |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service