Skip to Content
MilliporeSigma

F151-GS

Sigma-Aldrich

Feline Skeletal Muscle Cell Growth Supplement (50ml x 2)

Slide 1 of 1
Sign Into View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

form

liquid

Quality Level

shelf life

1 yr

manufacturer/tradename

Cell Applications, Inc

shipped in

dry ice

storage temp.

−20°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
F417-GSF411-GSCN151-GS
form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

shelf life

1 yr

shelf life

1 yr

shelf life

1 yr

shelf life

1 yr

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

General description

Added to Basal Medium to create Growth Medium

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service