All Photos(1)

F4519

Sigma-Aldrich

Sigma glass-fiber filters

retention >1.5 μm, circles diam. × thickness 9.0 cm × 0.65 mm

material

borosilicate glass

feature

speed medium

circles diam. × thickness

9.0 cm × 0.65 mm

pore size

>1.5 μm retention

basis weight

137 g/m2

Looking for similar products? Visit Product Comparison Guide

Application

For cell harvesting, receptor binding assays, RIA.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service