F9637

Sigma-Aldrich

Anti-Human IgA (α-chain specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

MDL number:

biological source

goat

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

application(s)

dot immunobinding: 1:64
immunoelectrophoresis: 1:8
particle immunofluorescence: 1:64

conjugate

FITC conjugate

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Specificity

No cross-reaction with Mouse and Rat IgG

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.01% thimerosal as a preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.