Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G1775

Sigma-Aldrich

Guillard′s (F/2) Marine Enrichment Basal Salt Mixture

powder, suitable for plant cell culture

Synonym(s):

F/2 Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

storage temp.

2-8°C

Application

Guillard′s (F/2) Marine Enrichment Basal Salt Mixture is composed of macro- and micronutrients. Guillard′s (F/2) Marine Enrichment Basal Salt Mixture may be used as a chemstock to support the reconstitution of Guillard′s (F/2) Marine Enrichment medium. Guillard′s (F/2) Marine Enrichment medium is a widely used enriched seawater medium for growing costal marine algae (diatoms).

Formula variant

With the macro- and micronutrients as described by Guillard (1975).

pictograms

Flame over circleCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 3 - Skin Corr. 1B

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service