G212180

Sigma-Aldrich

DMT-2′O-Methyl-rG(tac) Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):
DMT-2′O-Methyl-rG(tac) Amidite
Empirical Formula (Hill Notation):
C53H64N7O10P
Molecular Weight:
990.09
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

product line

Proligo Reagents

assay

≥98.0% (reversed phase HPLC)
≥99% (31P-NMR)

application(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

compatibility

configured for PerkinElmer
configured for Polygen

nucleoside profile

base: guanosine
base protecting group: TAC
2' protecting group: methyl
5' protecting group: DMT
deprotection: fast

storage temp.

−20°C

SMILES string

COC@@H1C@H(OP(OCCC#N)N(C(C)C)C(C)C)C@@H(COC(c2ccccc2)(c3ccc(OC)cc3)c4ccc(OC)cc4)OC@H1n5cnc6C(=O)NC(NC(=O)COc7ccc(cc7)C(C)(C)C)=Nc56

InChI

1S/C53H64N7O10P/c1-34(2)60(35(3)4)71(68-30-14-29-54)70-46-43(31-67-53(37-15-12-11-13-16-37,38-19-23-40(63-8)24-20-38)39-21-25-41(64-9)26-22-39)69-50(47(46)65-10)59-33-55-45-48(59)57-51(58-49(45)62)56-44(61)32-66-42-27-17-36(18-28-42)52(5,6)7/h11-13,15-28,33-35,43,46-47,50H,14,30-32H2,1-10H3,(H2,56,57,58,61,62)/t43-,46-,47-,50-,71?/m1/s1

InChI key

FEBFZGXNUHWEKM-BMBRRVPLSA-N

Looking for similar products? Visit Product Comparison Guide

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.