Skip to Content
MilliporeSigma

G8419

GW311616A

≥97% (HPLC), solid

Synonym(s):

(3S,3aS,6aR)-3-Isopropyl-1-(methanesulfonyl)-4-[4-(1-piperidinyl)-2(E)-butenoyl]perhydropyrrolo[3,2b]pyrrol-2(1H)-one hydrochloride

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H32ClN3O4S
CAS Number:
Molecular Weight:
433.99
UNSPSC Code:
12352200
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

Cl[H].CC(C)[C@H]1[C@H]2[C@H](CCN2C(=O)\C=C\CN3CCCCC3)N(C1=O)S(C)(=O)=O

assay

≥97% (HPLC)

form

solid

solubility

H2O: soluble 24 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

Potent, intracellular human neutrophil elastase (HNE, α-1-proteinase) inhibitor.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service