Documents

GBL204870

Grace Bio-Labs ProPlate® tray set

for Delrin® snap clip modules

NACRES:
NB.15

sterility

non-sterile

manufacturer/tradename

Grace Bio-Labs 204870

color

none

matrix active group

Non-treated surface treatment

Looking for similar products? Visit Product Comparison Guide

General description

holds up to 4 slides, includes cover, applicator, and 10 seal strips

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.