MilliporeSigma
All Photos(1)

Documents

H1020

Sigma-Aldrich

Heparin disaccharide II-S sodium salt

Sign Into View Organizational & Contract Pricing

Synonym(s):
α-ΔUA-[1→4]-GlcNS-6S
Linear Formula:
C12H16NO16S2Na3
CAS Number:
Molecular Weight:
563.35
MDL number:
PubChem Substance ID:

assay

≥98%

form

powder

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[Na].OC(COS(O)(=O)=O)C(OC1OC(=CC(O)C1O)C(O)=O)C(O)C(NS(O)(=O)=O)C=O

InChI

1S/C12H19NO16S2.Na.H/c14-2-4(13-30(21,22)23)8(17)10(6(16)3-27-31(24,25)26)29-12-9(18)5(15)1-7(28-12)11(19)20;;/h1-2,4-6,8-10,12-13,15-18H,3H2,(H,19,20)(H,21,22,23)(H,24,25,26);;

InChI key

DMHOLCPXAGVRLK-UHFFFAOYSA-N

Biochem/physiol Actions

Products of the digestion of heparin and heparan sulfate by various heparinases

Other Notes

In the following listings, δUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcN = D-glucosamine; Ac = Acetyl; NS, 2S, 6S, = N-sulfo, 2-sulfate and 6-sulfate respectively.

Preparation Note

Produced by the action of heparinase II

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service