H1388

Sigma-Aldrich

Human Serum

from clotted human male whole blood, sterile-filtered, (mycoplasma tested, virus tested)

MDL number:

biological source

human male whole blood (clotted)

sterility

sterile-filtered

impurities

(mycoplasma tested, virus tested)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Warning

Each donor is tested for and found non-reactive for Hepatitis B & C and non-reactive for Human Immunodeficiency Virus (HIV) antibody by ELISA. Nevertheless, products of human origin should be considered potentially infectious and handled accordingly.

Analysis Note

Endotoxin tested

Certificate of Analysis

Certificate of Origin

Hyo Joon Kim et al.
Clinical science (London, England : 1979), 130(2), 105-116 (2015-11-01)
Although apolipoprotein B100 (ApoB100) plays a key role in peripheral fat deposition, it is not considered a suitable therapeutic target in obesity. In the present study we describe a novel ApoB100 mimotope, peptide pB1, and the use of pB1-based vaccine-like...
Sangeeta Sule et al.
PloS one, 6(3), e17495-e17495 (2011-03-23)
Monocytes are a key component of the innate immune system involved in the regulation of the adaptive immune response. Previous studies have focused on apoptotic cell clearance abnormalities in systemic lupus erythematosus (SLE) monocytes. However, whether SLE monocytes might express...
Nathan P Wiederhold et al.
Antimicrobial agents and chemotherapy, 52(11), 4145-4148 (2008-09-17)
Pyrosequencing was compared to Sanger dideoxy sequencing to detect mutations in FKS1 responsible for reduced echinocandin susceptibility in Candida albicans. These methods were in complete agreement for 10 of 12 clinical isolates with elevated echinocandin MICs, supporting the potential feasibility...
Olívia Bacellar et al.
The Journal of infectious diseases, 200(1), 75-78 (2009-05-30)
Interleukin 17 (IL-17) plays a critical role in inflammation and autoimmunity. Very little is known about IL-17 in protozoa infection. Here, we show that lymphocytes obtained from patients with mucosal leishmaniasis and cutaneous leishmaniasis produce higher levels of IL-17 than...
Bonnie A Colleton et al.
Journal of virology, 83(12), 6288-6299 (2009-04-10)
Induction of an antigenically broad and vigorous primary T-cell immune response by myeloid dendritic cells (DC) in blood and tissues could be important for an effective prophylactic or therapeutic vaccine to human immunodeficiency virus type 1 (HIV-1). Here we show...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service