H6644

Sigma-Aldrich

Humidity chamber

overall size 9 in. × 4 in., trays size 8 in. × 2 1/2  in. , with lid

overall size

9 in. × 4 in.

trays size

8 in. × 2 1/2  in. , with lid

Other Notes

Tray with lid provides a constant humid environment for histochemical or immunohistochemical staining prodedures. The chamber is reusable and will hold six standard slides (1 in. × 3 in.). It is not designed to hold extremely hot or boiling water.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Rehemah Gwokyalya et al.
Archives of insect biochemistry and physiology, 101(4), e21588-e21588 (2019-06-11)
Boric acid (BA) is widely used in various industrial process and can be accessed to nontarget organisms. This study aimed to investigate the insecticidal effects of BA and its toxic activities with respect to immunologic and genotoxic effects using Galleria...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.