MilliporeSigma
All Photos(1)

H6757

Sigma-Aldrich

Methyl heptadecanoate

~95%

Synonym(s):
Methyl margarate, Heptadecanoic acid methyl ester
Linear Formula:
CH3(CH2)15COOCH3
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782681
EC Number:
MDL number:

assay

~95%

bp

152-153 °C/0.05 mmHg (lit.)

mp

29.8-30.3 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3

InChI key

HUEBIMLTDXKIPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service