Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

H7133

Heptafluorobutyric acid

Synonym(s):

Edman Reagent No. 3, HFBA, Perfluorobutyric acid

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CF3CF2CF2COOH
CAS Number:
Molecular Weight:
214.04
EC Number:
206-786-3
UNSPSC Code:
12352200
MDL number:
Beilstein/REAXYS Number:
1426882

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C4HF7O2/c5-2(6,1(12)13)3(7,8)4(9,10)11/h(H,12,13)

InChI key

YPJUNDFVDDCYIH-UHFFFAOYSA-N

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)F

vapor density

7 (vs air)

vapor pressure

~10 mmHg ( 25 °C)

refractive index

n20/D 1.3 (lit.)

bp

120 °C/755 mmHg (lit.)

density

1.645 g/mL at 25 °C (lit.)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrika Eriksson et al.
Environmental science and pollution research international, 20(11), 7940-7948 (2013-04-17)
Diet and drinking water are suggested to be major exposure pathways for perfluoroalkyl substances (PFASs). In this study, food items and water from Faroe Islands sampled in 2011/2012 were analyzed for 11 perfluoroalkyl carboxylic acids (PFCAs) and 4 perfluoroalkane sulfonic
Yuko Murakami et al.
Organic & biomolecular chemistry, 12(48), 9887-9894 (2014-10-31)
Desmosine-CH2, an analog of the elastic tissue degradation biomarker desmosine, can be regarded as a potential internal standard for precise quantification of desmosines by LC-MS/MS. In this study, the chemical synthesis of desmosine-CH2 was completed in 22% overall yield in
V Gellrich et al.
Chemosphere, 87(9), 1052-1056 (2012-03-07)
Perfluorinated compounds (PFCs) can be detected worldwide in both, soil and water. In order to study the leaching behavior of this heterogeneous group of compounds in soil, flow-through column experiments have been conducted. Ten perfluoro carboxylates and four perfluoro sulfonates
Merja R Häkkinen et al.
Journal of pharmaceutical and biomedical analysis, 45(4), 625-634 (2007-10-20)
A reversed phase liquid chromatography-electrospray ionization-tandem mass spectrometric method (RP-LC-ESI-MS/MS) was developed to separate and detect polyamines with minimal sample pre-treatment and without any derivatization. Prior to MS/MS analysis, a complete chromatographic separation of polyamines was achieved by a linear
Kaberi P Das et al.
Toxicological sciences : an official journal of the Society of Toxicology, 105(1), 173-181 (2008-05-31)
Perfluorobutyrate (PFBA) is a perfluoroalkyl acid (PFAA) found in the environment. Previous studies have indicated developmental toxicity of PFAAs (perfluorooctane sulfonate [PFOS] and perfluorooctanoate [PFOA]); the current study examines that of PFBA. PFBA/NH4(+) was given to timed-pregnant CD-1 mice by

Related Content

Product Information Sheet

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service