Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

H9886

Sigma-Aldrich

p-Hydroxybenzoate Hydroxylase from Pseudomonas sp.

lyophilized powder, ~20 units/mg solid

Synonym(s):

4-Hydroxybenzoate, NADPH:oxygen oxidoreductase (3-hydroxylating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

~20 units/mg solid

mol wt

55-~60 kDa

color

yellow

storage temp.

−20°C

Application

This enzyme is useful for enzymatic determination of choline esterase when coupled with protocatechuate 3, 4-dioxygenase.

Physical properties

Michaelis constants : 2.0 x 10-5M (p-Hydroxybenzoate), 4.0 x 10-5M (NADPH)
Structure : One mol of FAD per mol of enzyme
Inhibitors : Ag+, Hg++, PCMB, SDS
Optimum pH : 7.7 - 7.9
Optimum temperature : 35oC
pH Stability : pH 5.0 - 7.5 (25oC, 72hr)
Thermal stability : below 40oC (pH 6.0, 15min)

Unit Definition

One unit will hydroxylate 1.0 μmole of p-hydroxybenzoate to protocatechuate per min at pH 8.2 at 37 °C in the presence of NADPH.

Physical form

Contains mannitol and stabilizers

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsay J Cole et al.
Biochemistry, 44(22), 8047-8058 (2005-06-01)
p-Hydroxybenzoate hydroxylase (PHBH) is a homodimeric flavoprotein monooxygenase that catalyzes the hydroxylation of p-hydroxybenzoate to form 3,4-dihydroxybenzoate. Controlled catalysis is achieved by movement of the flavin and protein between three conformations, in, out, and open [Entsch, B., et al. (2005)
Adrie H Westphal et al.
The Journal of biological chemistry, 281(16), 11074-11081 (2006-02-24)
We have used the flavoenzyme p-hydroxybenzoate hydroxylase (PHBH) to illustrate that a strongly fluorescent donor label can communicate with the flavin via single-pair Förster resonance energy transfer (spFRET). The accessible Cys-116 of PHBH was labeled with two different fluorescent maleimides
Robert D Bach et al.
Journal of the American Chemical Society, 126(1), 127-142 (2004-01-08)
A model C-(4a)-flavinhydroperoxide (FlHOOH) is described that contains the tricyclic isoalloxazine moiety, the C-4a-hydroperoxide functionality, and a beta-hydroxyethyl group to model the effect of the 2'-OH group of the ribityl side chain of native FADHOOH. The electronic structures of this
Yue Cui et al.
Biosensors & bioelectronics, 22(11), 2754-2758 (2006-12-02)
This work reports the development of an amperometric glucose-6-phosphate biosensor by coimmobilizing p-hydroxybenzoate hydroxylase (HBH) and glucose-6-phosphate dehydrogenase (G6PDH) on a screen-printed electrode. The principle of the determination scheme is as follows: G6PDH catalyzes the specific dehydrogenation of glucose-6-phosphate by
Mariliz Ortiz-Maldonado et al.
Biochemistry, 43(6), 1569-1579 (2004-02-11)
Para-hydroxybenzoate hydroxylase is a flavoprotein monooxygenase that catalyzes a reaction in two parts: reduction of the enzyme cofactor, FAD, by NADPH in response to binding p-hydroxybenzoate to the enzyme, and oxidation of reduced FAD with oxygen to form a hydroperoxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service