Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA015827

Sigma-Aldrich

Anti-TRAK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 3 protein, Anti-Trafficking kinesin-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

HLYFSQSYGAFTGESLAAEIEGTMRKKLSLDEESSLFKQKAQQKRVFDTVRIANDTRGRSISFPALLPIPGSNRSSVIMTAKPFESGLQQTEDKSLLNQGSSSEEVAGSSQKMGQPGPSGDSDLATALHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRAK2(66008)

Immunogen

Trafficking kinesin-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Trafficking kinesin-binding protein 2 is a protein encoded by the TRAK2 gene in humans and belongs to TRAK family adaptor proteins. It mainly acts as a kinesin adaptor molecule, links to mitochondria to microtubule-based motors and is essential for mobility of axons and dendrites. This protein mainly interacts with dynein/dynactin and abundantly found in dendrites. This protein is important for the development of dendrites. Mitochondrial transport in neurons is regulated by this protein and helps in facilitating trafficking to cell surface.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73460

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam J Smith et al.
The Journal of biological chemistry, 281(37), 27216-27228 (2006-07-13)
Gamma-aminobutyric acid, type A (GABAA) receptor interacting factor-1 (GRIF-1) and N-acetylglucosamine transferase interacting protein (OIP) 106 are both members of a newly identified coiled-coil family of proteins. They are kinesin-associated proteins proposed to function as adaptors in the anterograde trafficking
Anatoly Grishin et al.
The Journal of biological chemistry, 281(40), 30104-30111 (2006-08-10)
To identify proteins that regulate potassium channel activity and expression, we performed functional screening of mammalian cDNA libraries in yeast that express the mammalian K(+) channel Kir2.1. Growth of Kir2.1-expressing yeast in media with low K(+) concentration is a function
Myrrhe van Spronsen et al.
Neuron, 77(3), 485-502 (2013-02-12)
In neurons, the distinct molecular composition of axons and dendrites is established through polarized targeting mechanisms, but it is currently unclear how nonpolarized cargoes, such as mitochondria, become uniformly distributed over these specialized neuronal compartments. Here, we show that TRAK
Omar Loss et al.
Journal of neuroscience research, 93(7), 1056-1066 (2015-02-06)
Neuronal function requires regulated anterograde and retrograde trafficking of mitochondria along microtubules by using the molecular motors kinesin and dynein. Previous work has established that trafficking kinesin proteins (TRAKs),TRAK1 and TRAK2, are kinesin adaptor proteins that link mitochondria to kinesin
Andrew F MacAskill et al.
Molecular and cellular neurosciences, 40(3), 301-312 (2008-12-24)
The transport of mitochondria to specific neuronal locations is critical to meet local cellular energy demands and for buffering intracellular calcium. A critical role for kinesin motor proteins in mitochondrial transport in neurons has been demonstrated. Currently however the molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service