HPA017163

Sigma-Aldrich

Anti-PLBD2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):
Anti-AC010178.40, Anti-LAMA-like protein 2, Anti-Lamina ancestor homolog 2, Anti-Putative phospholipase B-like 2 precursor, Anti-p76
Human Protein Atlas Number:

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

application(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GDWFSYDGSPRAQIFRRNQSLVQDMDSMVRLMRYNDFLHDPLSLCKACNPQPNGENAISARSDLNPANGSYPFQALRQRSHGGIDVKVTSMSLARILSLLAASGPTWDQVPPFQWSTS

conjugate

unconjugated

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... P76(196463)

General description

PLBD2 (phospholipase B domain containing 2) is a ~76kDa lysosomal localized protein. It consists of six N-glycosylation sites. It is identified as a mannose-6-phosphate protein. In prsence of Man6P (mannose 6-phosphate), PLBD2 directly interacts with cellular MPRs (Man6P receptors).

Immunogen

Putative phospholipase B-like 2 precursor recombinant protein epitope signature tag (PrEST)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72100.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Anaïs G Jensen et al.
The Biochemical journal, 402(3), 449-458 (2006-11-16)
Most soluble lysosomal proteins carry Man6P (mannose 6-phosphate), a specific carbohydrate marker that enables their binding to cellular MPRs (Man6P receptors) and their subsequent targeting towards the lysosome. This characteristic was exploited to identify novel soluble lysosomal proteins by proteomic...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.