HPA020920

Sigma-Aldrich

Anti-WNT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):
Anti-IRP protein, Anti-Int-1-related protein, Anti-Protein Wnt-2
Human Protein Atlas Number:

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

application(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

GSCTLRTCWLAMADFRKTGDYLWRKYNGAIQVVMNQDGTGFTVANERFKKPTKNDLVYFENSPDYCIRDREAGSLGTAGRVCNL

conjugate

unconjugated

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WNT2(7472)

General description

The gene WNT2 (wingless-type MMTV integration site family member 2) is mapped to human chromosome 7q31-33. It is a secreted glycoprotein.

Immunogen

Protein Wnt-2 Precursor recombinant protein epitope signature tag (PrEST)

Biochem/physiol Actions

WNT2 (wingless-type MMTV integration site family member 2) is important for cellular proliferation, invasion and angiogenesis, leading to tumor growth. In mouse model, it is needed for vascularization of the mouse placenta. WNT2 is up-regulated in esophageal, gastric, colorectal, breast, lung, cervical and malignant glioma. It is down-regulated in preeclampsia (PE). Mutations in WNT2 are linked with increased risk to autism. It is suggested to be involved in language development in humans suffering from autism.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74901.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

RIDADR

NONH for all modes of transport

WGK Germany

1

Certificate of Analysis
Certificate of Origin
Zhan Zhang et al.
Reproductive sciences (Thousand Oaks, Calif.), 20(8), 981-989 (2013-01-17)
The Wnt signaling pathway is a conserved pathway and plays a crucial role in regulating trophoblast functions. Abnormal expression of the Wnt pathway may result in the dysfunction of the trophoblast that can contribute to the pathogenesis of preeclampsia (PE)....
T H Wassink et al.
American journal of medical genetics, 105(5), 406-413 (2001-07-13)
We examined WNT2 as a candidate disease gene for autism for the following reasons. First, the WNT family of genes influences the development of numerous organs and systems, including the central nervous system. Second, WNT2 is located in the region...
Ping-I Lin et al.
Research in developmental disabilities, 33(5), 1533-1540 (2012-04-24)
Previous evidence suggests that language function is modulated by genetic variants on chromosome 7q31-36. However, it is unclear whether this region harbors loci that contribute to speech delay in autism. We previously reported that the WNT2 gene located on 7q31...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.