Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I106

Sigma-Aldrich

(S)-(+)-Ibuprofen

solid

Synonym(s):

(S)-(+)-2-(4-Isobutylphenyl)propionic acid, (S)-(+)-4-Isobutyl-α-methylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
206.28
Beilstein/REAXYS Number:
3590022
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

form

solid

optical activity

[α]26/D +54.46°, c = 0.6 in methanol(lit.)

color

white

mp

49-53 °C (lit.)

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.5 mg/mL
0.1 M NaOH: insoluble
methanol: soluble

SMILES string

CC(C)Cc1ccc(cc1)[C@H](C)C(O)=O

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/t10-/m0/s1

InChI key

HEFNNWSXXWATRW-JTQLQIEISA-N

Gene Information

human ... IL8RA(3577)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anti-inflammatory and analgesic. Active isomer of ibuprofen.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

R Mehvar et al.
Clinical chemistry, 34(3), 493-496 (1988-03-01)
This stereospecific "high-performance" liquid-chromatographic (HPLC) assay is suitable for pharmacokinetic studies of ibuprofen (IB). Very efficient extraction of the drug and internal standard, (+/-)-2-(4-benzoylphenyl)butyric acid, from plasma with isooctane/isopropanol (95/5, by vol) is followed by sequential reaction of the enantiomers
P Jeffrey et al.
The Journal of pharmacy and pharmacology, 43(10), 715-720 (1991-10-01)
The site of metabolic inversion of R(-)-ibuprofen to the pharmacologically active S(+)-enantiomer has been investigated using an array of in-situ rat perfused organ preparations allowing vascular perfusion (55-60 min) of the separate or combined intestine and liver. After addition of
R Mehvar et al.
Pharmaceutical research, 5(2), 76-79 (1988-02-01)
Equations describing plasma concentration-time courses of the individual enantiomers of chiral 2-arylpropionic acid nonsteroidal antiinflammatory drugs were derived from a general model. The model assumes first-order absorption and elimination of the enantiomers with presystemic and/or systemic R-to-S enantiomeric inversion. Utilizing
Surjeet Singh et al.
Bioorganic & medicinal chemistry letters, 17(13), 3706-3711 (2007-05-08)
The present study revealed the synergistic effect of boswellic acid mixture (BA) and glucosamine for anti-inflammatory and anti-arthritic activities in rats. Two studies were conducted, that is, acute anti-inflammatory by carrageenan edema and chronic anti-arthritic by Mycobacterium-induced developing arthritis. Five
Hong Su et al.
Bioorganic & medicinal chemistry letters, 21(12), 3578-3582 (2011-05-24)
Newly developed monofluoromethylation reaction provided access to various bioactive molecules with an interesting monofluoromethyl unit. An iridium-catalyzed asymmetric version was employed for large-scale methyl-monofluorination of widely used nonsteroidal anti-inflammatory drug ibuprofen (the active S isoform). The methyl-monofluorinated ibuprofen was found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service