I7758

Sigma-Aldrich

Anti-Human IgA (α-chain specific) antibody produced in goat

IgG fraction of antiserum, lyophilized powder

MDL number:
Pricing and availability is not currently available.

biological source

goat

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

packaging

vial of 2 mL lyophilized antiserum

application(s)

Ouchterlony double diffusion: 1:16

conjugate

unconjugated

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Kenneth A Kudsk et al.
The Journal of trauma, 64(2), 316-323 (2008-02-28)
Secretory immunoglobulin A (SIgA) is the specific immune antibacterial defense. Since pneumonia frequently complicates the course of trauma patients, we studied early airway immune responses after injury. Twelve severely injured, intubated (expected for >/=5 d) patients had tracheal and bilateral...
L R Haaheim et al.
Scandinavian journal of immunology, 43(1), 115-121 (1996-01-01)
The aim of this study was to investigate the epitope recognition pattern of La(SS-B) autoantibodies in sera from patients with Sjögren's syndrome (SS) and systemic lupus erythematosus (SLE) using overlapping synthetic decapeptides on solid phase. Eighty different decapeptides with five...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.