I8883

Sigma-Aldrich

Anti-Human Serum antibody produced in goat

IgG fraction of antiserum, whole antiserum, lyophilized powder

MDL number:

biological source

goat

antibody form

IgG fraction of antiserum
whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

packaging

vial of 2 mL lyophilized antiserum

conjugate

unconjugated

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.