All Photos(1)

I9634

Sigma-Aldrich

(−)-Isolongifolene

98% (GC)

Synonym(s):
(1R)-2,2,7,7-Tetramethyltricyclo[6.2.1.01.6]undec-5-ene
Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2207559
EC Number:
MDL number:

assay

98% (GC)

optical purity

optical purity: >90%

density

0.930 g/mL at 20 °C (lit.)

SMILES string

CC1(C)CCC=C2C(C)(C)[C@H]3CC[C@@]12C3

InChI

1S/C15H24/c1-13(2)8-5-6-12-14(3,4)11-7-9-15(12,13)10-11/h6,11H,5,7-10H2,1-4H3/t11-,15-/m0/s1

InChI key

CQUAYTJDLQBXCQ-NHYWBVRUSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Rienaecker, R. and Graefe, J
Angewandte Chemie (International Edition in English), 97, 348-348 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service