All Photos(3)

K2519

Sigma-Aldrich

Anti-KLF6 (171-184) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):
Anti-ZF9, Anti-BCD1, Anti-GBF, Anti-PAC1, Anti-COPEB, Anti-CPBP, Anti-Krueppel-like factor 6 isoform 1, Anti-ST12

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~32 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KLF6(1316)

Immunogen

synthetic peptide corresponding to amino acids 171-184 of human KLF6

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificate of Analysis

Certificate of Origin

Motokazu Koga et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(4), 383-391 (2014-06-21)
Pulmonary hypertension (PH) is associated with significant morbidity and mortality. Vasoactive intestinal peptide (VIP) and pituitary adenylyl cyclase activating peptide (PACAP) have pulmonary vasodilatory and positive inotropic effects via receptors VPAC1 and VPAC2, which possess a similar affinity for both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service