Skip to Content
MilliporeSigma

L1025

1-Linoleoyl-2-oleoyl-3-stearoyl-rac-glycerol

>99% (TLC), liquid

Synonym(s):

1-(cis,cis-9,12-Octadecadienoyl)-2-(cis-9-octadecenoyl)-3-octadecanoyl-rac-glycerol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C57H104O6
CAS Number:
Molecular Weight:
885.43
UNSPSC Code:
12352211
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>99% (TLC)

form

liquid

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCC\C=C\C\C=C\CCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC

General description

Used in the USP monograph for the analysis of sesame oil.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library