Skip to Content
MilliporeSigma

L3015

β-D-Lactopyranosylphenyl isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H25NO11S
CAS Number:
Molecular Weight:
475.47
UNSPSC Code:
12352201
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

form

powder

suitability

suitable for preparation of neoglycoproteins

storage temp.

2-8°C

SMILES string

OCC1OC(OC2C(CO)OC(Oc3ccc(cc3)N=C=S)C(O)C2O)C(O)C(O)C1O

InChI

1S/C19H25NO11S/c21-5-10-12(23)13(24)15(26)19(29-10)31-17-11(6-22)30-18(16(27)14(17)25)28-9-3-1-8(2-4-9)20-7-32/h1-4,10-19,21-27H,5-6H2

InChI key

NGHKRXUEFMEWTE-UHFFFAOYSA-N



pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library