All Photos(1)
L5149
Sigma-Aldrich
Labels, phosphorescent
4 × 6 inch sheets
packaging
pkg of 10 sheets
manufacturer/tradename
Diversified Biotech RAD-10
L × W
10.1 cm × 15.25 cm
Related Categories
General description
Label sheets for autoradiograms. Identifying marks are written on the label with an ordinary black pen, then affixed to a membrane or gel (the label is self-adhesive). A clear negative image of the markings is transferred to the autoradiogram, regardless of the length of the exposure.
Packaging
Package contains ten 4 × 6 inch sheets, which may be cut to size.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).