All Photos(1)

M7888

Sigma-Aldrich

Muristerone A

≥90%

Synonym(s):
MurA, 2β,3β,5β,11α,14α,20R,22R-Hepta­hydroxy­cholest-7-en-6-one
Linear Formula:
C27H44O8
MDL number:
PubChem Substance ID:

assay

≥90%

storage temp.

−20°C

SMILES string

CC(C)CC[C@@H](O)[C@](C)(O)C1CC[C@@]2(O)C3=CC(=O)[C@]4(O)C[C@@H](O)[C@@H](O)C[C@]4(C)C3[C@H](O)C[C@]12C

InChI

1S/C27H44O8/c1-14(2)6-7-20(31)25(5,33)19-8-9-26(34)15-10-21(32)27(35)13-17(29)16(28)11-24(27,4)22(15)18(30)12-23(19,26)3/h10,14,16-20,22,28-31,33-35H,6-9,11-13H2,1-5H3/t16-,17+,18+,19?,20+,22?,23+,24+,25+,26+,27+/m0/s1

InChI key

LRJUYAVTHIEHAI-WRENIRACSA-N

Biochem/physiol Actions

Muristerone A is a native phytosteroid known to have insecticidal properties. Induces apoptosis in cell transfected with wild-type Bax and ecdysone-inducible gene expression systems in mammalian cells and transgenic animals.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service