MilliporeSigma
All Photos(1)

Documents

M9513

Sigma-Aldrich

N-Methyl-bis(trifluoroacetamide)

~95% (capillary GC)

Sign Into View Organizational & Contract Pricing

Synonym(s):
MBTFA
Linear Formula:
CF3CON(CH3)COCF3
CAS Number:
Molecular Weight:
223.07
Beilstein/REAXYS Number:
1878402
EC Number:
MDL number:
PubChem Substance ID:

assay

~95% (capillary GC)

refractive index

n20/D 1.346 (lit.)

bp

121-122 °C (lit.)

density

1.547 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

InChI

1S/C5H3F6NO2/c1-12(2(13)4(6,7)8)3(14)5(9,10)11/h1H3

InChI key

AWGBWLXGUPTXHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for introducing trifluoroacetyl group.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

N Pizarro et al.
Journal of pharmaceutical and biomedical analysis, 21(4), 739-747 (2000-03-04)
We developed a fast and sensitive method for identification and quantification of plasma concentrations of amphetamine using gas chromatography with mass spectrometry detection (GC-MS). Amphetamine-d8 served as internal standard. The method involves a single extraction procedure and an easy treatment
Caroline Raeppel et al.
Journal of chromatography. A, 1218(44), 8123-8129 (2011-10-04)
This work presents a new method to analyse simultaneously by GC-MS 31 pesticides from different chemical classes (2,4 D, 2,4 MCPA, alphacypermethrin, bifenthrin, bromoxynil, buprofezin, carbaryl, carbofuran, clopyralid, cyprodinil, deltamethrin dicamba, dichlobenil, dichlorprop, diflufenican, diuron, fenoxaprop, flazasulfuron, fluroxypyr, ioxynil, isoxaben
Kathleen A Vermeersch et al.
PloS one, 10(2), e0118262-e0118262 (2015-02-18)
Recently, multicellular spheroids were isolated from a well-established epithelial ovarian cancer cell line, OVCAR-3, and were propagated in vitro. These spheroid-derived cells displayed numerous hallmarks of cancer stem cells, which are chemo- and radioresistant cells thought to be a significant
Devin H Currie et al.
BMC systems biology, 9, 30-30 (2015-06-27)
Thermoanaerobacterium saccharolyticum is a hemicellulose-degrading thermophilic anaerobe that was previously engineered to produce ethanol at high yield. A major project was undertaken to develop this organism into an industrial biocatalyst, but the lack of genome information and resources were recognized
Thomas B Kakule et al.
Journal of the American Chemical Society, 136(51), 17882-17890 (2014-12-02)
The programming of the fungal polyketide synthase (PKS) is quite complex, with a simple domain architecture leading to elaborate products. An additional level of complexity has been found within PKS-based pathways where the PKS is fused to a single module

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service