Skip to Content
MilliporeSigma

N0257

2-Nitrophenyl N-acetyl-α-D-galactosaminide

Synonym(s):

o-Nitrophenyl 2-acetamido-2-deoxy-α-D-galactopyranoside

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-5-3-2-4-8(9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)

SMILES string

CC(=O)NC1C(O)C(O)C(CO)OC1Oc2ccccc2N(=O)=O

InChI key

PXMQUEGJJUADKD-UHFFFAOYSA-N

form

solid

storage temp.

−20°C

comparable product

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean H White et al.
Journal of neurophysiology, 112(2), 446-462 (2014-04-18)
Nicotinic receptors form a diverse group of ligand-gated ionotropic receptors with roles in both synaptic transmission and the control of excitability. In the bag cell neurons of Aplysia, acetylcholine activates an ionotropic receptor, which passes inward current to produce a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service